Fipronil ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
5-Amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-3-cyano-4-(trifluoromethyl)sulphinylpyrazole, 5-Amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-(trifluoromethyl)sulphinylpyrazole-3-carbonitrile
F0822-1G
F0822-5G
TCIAF0822-1GEA
81
EUR
TCIAF0822-1G
TCIAF0822-5G
Fipronil ≥97.0% (by GC)
Fipronil
Formula:
C₁₂H₄Cl₂F₆N₄OS Peso molecolare: 437,14 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00867812 CAS Number: 120068-37-3 UN: 2811 ADR: 6.1,III Merck Index: 14,04085 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 196.0 to 205.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...