Stampa…

3-Formyl-2-thiopheneboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Formylthiophene-2-boronic acid, (3-Formylthiophen-2-yl)boronic acid

F0801-1G F0801-5G
TCIAF0801-1GEA 110 EUR
TCIAF0801-1G TCIAF0801-5G
3-Formyl-2-thiopheneboronic acid (contains varying amounts of Anhydride)
3-Formyl-2-thiopheneboronic acid
Formula: C₅H₅BO₃S
Peso molecolare: 155,97 g/mol
MDL Number: MFCD01075679
CAS Number: 17303-83-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77d83606d6566dfd3f3ae92a1976b11e58514df8&vid=77d83606d6566dfd3f3ae92a1976b11e58514df8&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (Neutralization titration) 97.0 to 113.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR