Stampa…

Flupirtine maleate salt ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: Ethyl (2-amino-6-((4-fluorobenzyl)amino)pyridin-3-yl)carbamate maleate, 2-Amino-3-(ethoxycarbonylamino)-6-(4-fluorobenzylamino)pyridine Maleate Salt, Flupirtine Maleate

F0736-1G F0736-100MG
TCIAF0736-1GEA 298 EUR
TCIAF0736-1G TCIAF0736-100MG
Flupirtine maleate salt ≥98.0% (by HPLC, titration analysis)
Flupirtine maleate salt
Formula: C₁₅H₁₇FN₄O₂
Peso molecolare: 420.40 g/mol
Punto di fusione: 177°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00941415
CAS Number: 75507-68-5
EINECS: 278-225-0
Merck Index: 14,04190

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR