Stampa…

6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

F0735-25G F0735-5G
TCIAF0735-25GEA 212 EUR
TCIAF0735-25G TCIAF0735-5G
6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole ≥98.0% (by GC, titration analysis)
6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole
Formula: C₁₂H₁₃FN₂O
Peso molecolare: 220,24 g/mol
Punto di fusione: 306°C
MDL Number: MFCD03236724
CAS Number: 84163-77-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR