Stampa…

4-Fluoro-4'-hydroxybiphenyl ≥98.0% (by GC)

Fornitore: TCI
Attenzione

F0704-25G F0704-5G
TCIAF0704-25GEA 198 EUR
TCIAF0704-25G TCIAF0704-5G
4-Fluoro-4'-hydroxybiphenyl ≥98.0% (by GC)
4-Fluoro-4'-hydroxybiphenyl
Formula: C₁₂H₉FO
MDL Number: MFCD01830385
CAS Number: 324-94-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 170.0 to 174.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR