Stampa…

Fenofibrate ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionic Acid Isopropyl Ester, Isopropyl 2-(4-(4-chlorobenzoyl)phenoxy)-2-methylpropanoate, Isopropyl 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionate

F0674-25G F0674-5G F0674-100G
TCIAF0674-25GEA 134 EUR
TCIAF0674-25G TCIAF0674-5G TCIAF0674-100G
Fenofibrate ≥98.0% (by GC)
Fenofibrate
Formula: C₂₀H₂₁ClO₄
Peso molecolare: 360,83 g/mol
MDL Number: MFCD00133314
CAS Number: 49562-28-9
EINECS: 256-376-3
Merck Index: 14,03978

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 79.0 to 83.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR