Stampa…

Fluocinolone acetonide ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 6alpha,9alpha-Difluoro-11beta,16alpha,17alpha,21-tetrahydroxy-1,4-pregnadiene-3,20-dione 16,17-Acetonide

F0657-1G F0657-5G
TCIAF0657-1GEA 164 EUR
TCIAF0657-1G TCIAF0657-5G
Fluocinolone acetonide ≥98.0% (by HPLC)
Fluocinolone acetonide
Formula: C₂₄H₃₀F₂O₆
Peso molecolare: 452,49 g/mol
Punto di fusione: 266°C
MDL Number: MFCD00010525
CAS Number: 67-73-2
EINECS: 200-668-5
Merck Index: 14,04150

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D +100.0 to +106.0 deg (C=1, methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR