Stampa…

2-Fluoroadenine ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 6-Amino-2-fluoropurine, 6-Amino-2-fluoro-7H-purine, 2-Fluoro-7H-purin-6-amine

F0647-1G F0647-200MG
TCIAF0647-1GEA 204 EUR
TCIAF0647-1G TCIAF0647-200MG
2-Fluoroadenine ≥98.0% (by HPLC, titration analysis)
2-Fluoroadenine
Formula: C₅H₄FN₅
Peso molecolare: 153,12 g/mol
MDL Number: MFCD01632749
CAS Number: 700-49-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR