Stampa…

3-Fluoro-4-idrossibenzonitrile ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

F0638-25G F0638-5G
TCIAF0638-25GEA 341 EUR
TCIAF0638-25G TCIAF0638-5G
3-Fluoro-4-idrossibenzonitrile ≥98.0% (by GC, titration analysis)
3-Fluoro-4-idrossibenzonitrile
Formula: C₇H₄FNO
Peso molecolare: 137,11 g/mol
MDL Number: MFCD00215834
CAS Number: 405-04-9
UN: 3439
ADR: 6.1,III

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 134.0 to 138.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR