5-Fluoro-1-(tetrahydro-2-furfuryl)uracil ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI
|
Pericolo
|
Synonyms:
Ftorafur, Tegafur, 5-Fluoro-1-(tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione
Formula:
C₈H₉FN₂O₃ |
MDL Number:
MFCD00012351 CAS Number: 17902-23-7 EINECS: 241-846-2 UN: 2811 ADR: 6.1,II Merck Index: 14,09112 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 167.0 to 171.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...