N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-threonine ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
Synonyms:
(2R,3S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxybutanoic acid, N-Fmoc-D-threonine, Fmoc-D-Thr-OH
Formula:
C₁₉H₁₉NO₅ Peso molecolare: 341.36 g/mol |
MDL Number:
MFCD00153365 CAS Number: 157355-81-2 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % (calcd.on anh.substance) |
Specific rotation [a]20/D | +14.0 to +17.0 deg (C=1, DMF) |
Solubility in Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...