Stampa…

Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-lysine hydrochloride ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: N2-Fmoc-D-lysine Hydrochloride, Fmoc-D-Lys-OH.HCl, Nɑ-Fmoc-D-lysine Hydrochloride

F0604-1G F0604-5G
TCIAF0604-1GEA 74 EUR
TCIAF0604-1G TCIAF0604-5G
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-lysine hydrochloride ≥98.0% (by HPLC)
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-lysine hydrochloride
Formula: C₂₁H₂₄N₂O₄
Peso molecolare: 404,89 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00190888
CAS Number: 201002-47-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 90.0 %
Specific rotation [a]20/D +9.0 to +13.0 deg (C=1, DMF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR