Stampa…

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-aspartic acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: Fmoc-D-Asp-OH, N-Fmoc-D-aspartic Acid

F0592-1G F0592-5G
TCIAF0592-1GEA 16 EUR
TCIAF0592-1G TCIAF0592-5G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-aspartic acid ≥98.0% (by HPLC, titration analysis)
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-aspartic acid
Formula: C₁₉H₁₇NO₆
Peso molecolare: 355,35 g/mol
MDL Number: MFCD01318740
CAS Number: 136083-57-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +27.0 to +31.0 deg (C=1, DMF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR