N-(4-Fluorophenyl)formamide ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
4'-Fluoroformanilide, 1-Fluoro-4-formamidobenzene
F0546-5G
F0546-1G
TCIAF0546-5GEA
138
EUR
TCIAF0546-5G
TCIAF0546-1G
N-(4-Fluorophenyl)formamide ≥98.0% (by GC)
N-(4-Fluorophenyl)formamide
Formula:
C₇H₆FNO Peso molecolare: 139,13 g/mol |
CAS Number:
459-25-6 |
Risultati del test di specificazione
Appearance | White to Gray to Red powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 66.0 to 70.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...