4-Fluorobenzhydrazide ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4-Fluorobenzoic hydrazide, 4-Fluorobenzene-1-carbohydrazide, 4-Fluorobenzoic acid hydrazide, 4-Fluorobenzohydrazide
F0540-25G
F0540-5G
TCIAF0540-25GEA
422
EUR
TCIAF0540-25G
TCIAF0540-5G
4-Fluorobenzhydrazide ≥98.0% (by GC, titration analysis)
4-Fluorobenzhydrazide
Formula:
C₇H₇FN₂O Peso molecolare: 154,14 g/mol |
MDL Number:
MFCD00060562 CAS Number: 456-06-4 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Bromination Method) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...