Stampa…

(1-Fluorovinyl)methyldiphenylsilane ≥96.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-Fluoro-1-(methyldiphenylsilyl)ethylene

F0518-1G F0518-5G
TCIAF0518-1GEA 147 EUR
TCIAF0518-1G TCIAF0518-5G
(1-Fluorovinyl)methyldiphenylsilane ≥96.0% (by GC)
(1-Fluorovinyl)methyldiphenylsilane
Formula: C₁₅H₁₅FSi
Peso molecolare: 242,37 g/mol
CAS Number: 257610-49-4

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR