Stampa…

Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-γ-trityl-L-asparagine ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: Fmoc-Asn(Trt)-OH, Nɑ-Fmoc-Nγ-trityl-L-asparagine

F0508-1G F0508-25G F0508-5G
TCIAF0508-1GEA 16 EUR
TCIAF0508-1G TCIAF0508-25G TCIAF0508-5G
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-γ-trityl-L-asparagine ≥98.0% (by HPLC, titration analysis)
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-gamma-trityl-L-asparagine
Formula: C₃₈H₃₂N₂O₅
Peso molecolare: 596,68 g/mol
MDL Number: MFCD00077049
CAS Number: 132388-59-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -17.0 to -21.0 deg (C=1, DMF)
Solubility in N,N-DMF almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR