Stampa…

N-(9-Fluorenylmethoxycarbonyl)-O-tert-butyl-L-threonine ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI

Synonyms: Fmoc-Thr(tBu)-OH, Nalpha-Fmoc-O-tert-butyl-L-threonine, O-tert-Butyl-Nalpha-Fmoc-L-threonine, FMOC-O-tert-Butyl-L-threonine

F0505-1G F0505-25G F0505-5G
TCIAF0505-1GEA 16 EUR
TCIAF0505-1G TCIAF0505-25G TCIAF0505-5G
N-(9-Fluorenylmethoxycarbonyl)-O-tert-butyl-L-threonine ≥98.0% (by HPLC, titration analysis)
N-(9-Fluorenylmethoxycarbonyl)-O-tert-butyl-L-threonine
Formula: C₂₃H₂₇NO₅
Peso molecolare: 397.5 g/mol
MDL Number: MFCD00077075
CAS Number: 71989-35-0
EINECS: 276-261-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 131.0 to 135.0 deg-C
Specific rotation [a]20/D +14.0 to +17.0 deg (C=1, EtOAc)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR