Stampa…

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-tyrosine ≥95.0% (by HPLC, titration analysis)

Fornitore: TCI

Synonyms: Fmoc-Tyr-OH, N-Fmoc-L-tyrosine

F0456-1G F0456-5G
TCIAF0456-1GEA 32 EUR
TCIAF0456-1G TCIAF0456-5G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-tyrosine ≥95.0% (by HPLC, titration analysis)
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-tyrosine
Formula: C₂₄H₂₁NO₅
Peso molecolare: 403.43 g/mol
MDL Number: MFCD00134890
CAS Number: 92954-90-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 95.0 %
Specific rotation [a]20/D -20.0 to -24.0 deg (C=1, DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR