Stampa…

Ethyl (4-fluorobenzoyl)acetate ≥98.0% (by GC, titration analysis)

Fornitore: TCI

Synonyms: Ethyl 3-(4-Fluorophenyl)-3-oxopropionate, 3-(4-Fluorophenyl)-3-oxopropionic Acid Ethyl Ester, (4-Fluorobenzoyl)acetic Acid Ethyl Ester, Ethyl 3-(4-fluorophenyl)-3-oxopropanoate

F0435-1G F0435-25G F0435-5G
TCIAF0435-1GEA 48 EUR
TCIAF0435-1G TCIAF0435-25G TCIAF0435-5G
Ethyl (4-fluorobenzoyl)acetate ≥98.0% (by GC, titration analysis)
Ethyl (4-fluorobenzoyl)acetate
Formula: C₁₁H₁₁FO₃
Peso molecolare: 210.20 g/mol
Punto di ebollizione: 120°C
MDL Number: MFCD03093631
CAS Number: 1999-00-4

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 % (total of isomer)
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR