Stampa…

N-Formylglycine ethyl ester ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: N-FormylglycineEthylEster, Formamidoacetic Acid Ethyl Ester, For-Gly-OEt, Ethyl Formamidoacetate

F0421-25G F0421-5G
TCIAF0421-25GEA 163 EUR
TCIAF0421-25G TCIAF0421-5G
N-Formylglycine ethyl ester ≥98.0% (by GC)
N-Formylglycine ethyl ester
Formula: C₅H₉NO₃
Peso molecolare: 131,13 g/mol
MDL Number: MFCD00014125
CAS Number: 3154-51-6
EINECS: 221-596-0

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR