Stampa…

3-Fluoro-4-hydroxybenzoic acid, idrato ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

F0413-1G F0413-5G
TCIAF0413-1GEA 31 EUR
TCIAF0413-1G TCIAF0413-5G
3-Fluoro-4-hydroxybenzoic acid, idrato ≥98.0% (by GC, titration analysis)
3-Fluoro-4-hydroxybenzoic acid
Formula: C₇H₅FO₃
Peso molecolare: 156,11 g/mol
MDL Number: MFCD02094741
CAS Number: 350-29-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Melting point 161.0 to 165.0 deg-C
Water max. 10.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR