Stampa…

4-Formyl-trans-stilbene ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: trans-4-Stilbenecarboxaldehyde

F0412-5G F0412-25G
TCIAF0412-5GEA 110 EUR
TCIAF0412-5G TCIAF0412-25G
4-Formyl-trans-stilbene ≥98.0% (by GC)
4-Formyl-trans-stilbene
Formula: C₁₅H₁₂O
Peso molecolare: 208,26 g/mol
MDL Number: MFCD00011580
CAS Number: 40200-69-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 112.0 to 115.0 deg-C
Solubility in hot EtOH almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR