Stampa…

2-Phenylethyl formate ≥95.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Phenethyl Formate, Formic Acid 2-Phenylethyl Ester, Formic Acid Phenethyl Ester

F0363-25G F0363-500G
TCIAF0363-25GEA 16 EUR
TCIAF0363-25G TCIAF0363-500G
2-Phenylethyl formate ≥95.0% (by GC)
2-Phenylethyl formate
Formula: C₉H₁₀O₂
Peso molecolare: 150,18 g/mol
Punto di ebollizione: 117°C
Flash Pt: 103°C
MDL Number: MFCD00021046
CAS Number: 104-62-1
EINECS: 203-220-7

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR