4-(2-Furyl)-3-buten-2-one ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₈H₈O₂ Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00039566 CAS Number: 623-15-4 EINECS: 210-774-3 |
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to lump |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...