Stampa…

Butyl fluoroacetate ≥97.0% (by GC)

Fornitore: TCI
Pericolo

Synonyms: Fluoroacetic Acid Butyl Ester

F0215-25ML F0215-5ML
TCIAF0215-25MLEA 299 EUR
TCIAF0215-25ML TCIAF0215-5ML
Butyl fluoroacetate ≥97.0% (by GC)
Butyl fluoroacetate
Formula: C₆H₁₁FO₂
Punto di ebollizione: 154°C
MDL Number: MFCD00042194
CAS Number: 371-49-3
EINECS: 206-739-7
UN: 2929
ADR: 6.1,II

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Specific gravity (20/20) 1.0200 to 1.0230
Refractive index n20/D 1.3940 to 1.3970


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR