Stampa…

Fluorescamine for HPLC labeling

Fornitore: TCI EUROPE

Synonyms: Fluram, 4-Phenyl-3H,3'H-spiro[furan-2,1'-isobenzofuran]-3,3'-dione, 4-Phenylspiro[furan-2(3H),1'-phthalan]-3,3'-dione

F0192-1G F0192-100MG
TCIAF0192-1GEA 396 EUR
TCIAF0192-1G TCIAF0192-100MG
Fluorescamine for HPLC labeling
Fluorescamine
Formula: C₁₇H₁₀O₄
Peso molecolare: 278,26 g/mol
MDL Number: MFCD00005928
CAS Number: 38183-12-9
EINECS: 253-814-5
Merck Index: 14,04158

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Melting point 155.0 to 159.0 deg-C
Lambda max. 233.0 to 235.0 nm (Diethyl ether)
Molar extinction coefficient min. 26,000 (Diethyl ether, 233.0 to 235.0 nm)
Fluorescence test Emit Fluorescence in the presence of Glycine

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR