Stampa…

Furosemide ≥99.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: 5-(Aminosulphonyl)-4-chloro-2-[(2-furanylmethyl)amino]benzoic acid, 4-Chloro-N-furfuryl-5-sulphamoylanthranilic acid

F0182-25G F0182-5G
TCIAF0182-25GEA 182 EUR
TCIAF0182-25G TCIAF0182-5G
Furosemide ≥99.0% (by titrimetric analysis)
Furosemide
Formula: C₁₂H₁₁ClN₂O₅S
Peso molecolare: 330,74 g/mol
MDL Number: MFCD00010549
CAS Number: 54-31-9
EINECS: 200-203-6
Merck Index: 14,04309

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 99.0 %
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR