Stampa…

ɑ-Furil dioxime ≥97.0% (W)

Fornitore: TCI EUROPE

F0079-1G F0079-5G
TCIAF0079-1GEA 312 EUR
TCIAF0079-1G TCIAF0079-5G
ɑ-Furil dioxime ≥97.0% (W)
alpha-Furil dioxime
Formula: C₁₀H₈N₂O₄
Peso molecolare: 220,18 g/mol
MDL Number: MFCD00075315
CAS Number: 23789-34-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light red powder to crystal
Purity (Gravimetric Method) min. 97.0 %
Solubility in hot EtOH almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR