Stampa…

Isoamyl formate ≥95.0% (by GC)

Fornitore: TCI
Pericolo

Synonyms: Formic Acid Isoamyl Ester

F0054-25ML F0054-500ML
TCIAF0054-25MLEA 16 EUR
TCIAF0054-25ML TCIAF0054-500ML
Isoamyl formate ≥95.0% (by GC)
Isoamyl formate
Formula: C₆H₁₂O₂
Punto di ebollizione: 124°C
Flash Pt: 25°C
MDL Number: MFCD00021049
CAS Number: 110-45-2
EINECS: 203-769-2
UN: 1109
ADR: 3,III
Merck Index: 14,05119

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR