(S)-Ethyl-2-((S)-4-methyl-2,5-dioxooxazolidin-3-yl)-4-phenylbutanoate ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Pericolo
|
E1423-1G
E1423-5G
TCIAE1423-1GEA
19
EUR
TCIAE1423-1G
TCIAE1423-5G
(S)-Ethyl-2-((S)-4-methyl-2,5-dioxooxazolidin-3-yl)-4-phenylbutanoate ≥98.0% (by HPLC, titration analysis)
(S)-Ethyl-2-((S)-4-methyl-2,5-dioxooxazolidin-3-yl)-4-phenylbutanoate
Formula:
C₁₆H₁₉NO₅ Punto di ebollizione: 345°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00272296 CAS Number: 84793-24-8 EINECS: 430-360-8 |
Risultati del test di specificazione
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 69.0 to 73.0 deg-C |
Specific rotation [a]20/D | 34.0 to 38.0 deg (C=1, CHCL3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...