Stampa…

1-Ethylcyclohexyl methacrylate ≥98.0% (by GC) stabilizzato

Fornitore: TCI EUROPE

Synonyms: Methacrylic acid 1-Ethylcyclohexyl ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

E1407-1G E1407-5G
TCIAE1407-1GEA 42 EUR
TCIAE1407-1G TCIAE1407-5G
1-Ethylcyclohexyl methacrylate ≥98.0% (by GC) stabilizzato
1-Ethylcyclohexyl methacrylate
Formula: C₁₂H₂₀O₂
Peso molecolare: 196,29 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD30470335
CAS Number: 274248-09-8

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR