4-Ethoxyphenyl trans-4-Propylcyclohexanecarboxylate ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Trans-4-ethoxyphenyl 4-propylcyclohexanecarboxylate, trans-4-Propylcyclohexanecarboxylic acid 4-Ethoxyphenyl ester
E1395-25G
E1395-100G
TCIAE1395-25GEA
144
EUR
TCIAE1395-25G
TCIAE1395-100G
4-Ethoxyphenyl trans-4-Propylcyclohexanecarboxylate ≥98.0% (by GC)
4-Ethoxyphenyl trans-4-Propylcyclohexanecarboxylate
Formula:
C₁₈H₂₆O₃ Peso molecolare: 290,40 g/mol |
MDL Number:
MFCD11053417 CAS Number: 67589-39-3 EINECS: 266-744-5 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | max. 98.0 % |
Mesomorphic range | 33.0 to 78.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...