Ethyl 2-(3-Formyl-4-hydroxyphenyl)-4-methylthiazole-5-carboxylate ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
Ethyl 2-(3-formyl-4-hydroxyphenyl)-4-methylthiazole-5-carboxylate
E1394-5G
E1394-25G
TCIAE1394-5GEA
33
EUR
TCIAE1394-5G
TCIAE1394-25G
Ethyl 2-(3-Formyl-4-hydroxyphenyl)-4-methylthiazole-5-carboxylate ≥98.0% (by GC, titration analysis)
Ethyl 2-(3-Formyl-4-hydroxyphenyl)-4-methylthiazole-5-carboxylate
Formula:
C₁₄H₁₃NO₄S Peso molecolare: 291,32 g/mol |
MDL Number:
MFCD13194810 CAS Number: 161798-01-2 |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 113.0 to 117.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...