1-Ethylcyclopentyl methacrylate ≥98.0% (by GC) stabilizzato
Fornitore: TCI
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
Formula:
C₁₁H₁₈O₂ Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD23703079 CAS Number: 266308-58-1 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...