Stampa…

Ethyl-5-methyl-1H-imidazole-4-carboxylate ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: Ethyl 4-Methyl-1H-imidazole-5-carboxylate, 4-Methyl-1H-imidazole-5-carboxylic acid ethyl ester, Ethyl 4-methyl-5-imidazolecarboxylate, Ethyl 5-methyl-1H-imidazole-4-carboxylate

E1386-5G E1386-25G
TCIAE1386-5GEA 31 EUR
TCIAE1386-5G TCIAE1386-25G
Ethyl-5-methyl-1H-imidazole-4-carboxylate ≥98.0% (by HPLC)
Ethyl-5-methyl-1H-imidazole-4-carboxylate
Formula: C₇H₁₀N₂O₂
Peso molecolare: 154,1665 g/mol
MDL Number: MFCD00005199
CAS Number: 51605-32-4
EINECS: 257-315-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 203.0 to 207.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR