Stampa…

N,N'-(Ethane-1,2-diylidene)bis(2,4,6-trimethylaniline) ≥98.0% (by GC, total nitrogen)

Fornitore: TCI EUROPE

Synonyms: N,N'-Bis(2,4,6-trimethylphenyl)ethane-1,2-diimine

E1383-1G E1383-200MG
TCIAE1383-1GEA 185 EUR
TCIAE1383-1G TCIAE1383-200MG
N,N'-(Ethane-1,2-diylidene)bis(2,4,6-trimethylaniline) ≥98.0% (by GC, total nitrogen)
N,N'-(Ethane-1,2-diylidene)bis(2,4,6-trimethylaniline)
Formula: C₂₀H₂₄N₂
Peso molecolare: 292,43 g/mol
MDL Number: MFCD06669905
CAS Number: 56222-36-7

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 155.0 to 159.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR