Stampa…

Ethyl 2-(trans-4-Aminocyclohexyl)acetate hydrochloride ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: trans-2-(4-Aminocyclohexyl)acetic acid Ethyl ester hydrochloride, Ethyl 2-(trans-4-aminocyclohexyl)acetate hydrochloride

E1374-1G E1374-5G
TCIAE1374-1GEA 92 EUR
TCIAE1374-1G TCIAE1374-5G
Ethyl 2-(trans-4-Aminocyclohexyl)acetate hydrochloride ≥98.0% (by titrimetric analysis)
Ethyl 2-(trans-4-Aminocyclohexyl)acetate hydrochloride
Formula: C₁₀H₁₉NO₂·HCl
Peso molecolare: 221,73 g/mol
MDL Number: MFCD16660366
CAS Number: 76308-26-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 175.0 to 179.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR