trans-1-Ethoxy-4-(4-pentylcyclohexyl)benzene ≥98.0% (by GC)
Fornitore: TCI
Formula:
C₁₉H₃₀O |
MDL Number:
MFCD06658196 CAS Number: 84540-32-9 EINECS: 283-125-5 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 51.0 to 55.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...