Stampa…

Ethyl 2,3-Dicyanopropionate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Ethyl 2,3-dicyanopropanoate, 2,3-Dicyanopropanoic acid Ethyl ester, 2,3-Dicyanopropionic acid Ethyl ester, Ethyl 2,3-Dicyanopropanate

E1366-25G E1366-5G
TCIAE1366-25GEA 142 EUR
TCIAE1366-25G TCIAE1366-5G
Ethyl 2,3-Dicyanopropionate ≥98.0% (by GC)
Ethyl 2,3-Dicyanopropionate
Formula: C₇H₈N₂O₂
Peso molecolare: 152,15 g/mol
Punto di ebollizione: 140°C
MDL Number: MFCD08061844
CAS Number: 40497-11-8

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Acid Value max. 1.0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR