Stampa…

Estradiol Dipropionate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (17beta)-Estra-1(10),2,4-triene-3,17-diyl Dipropanoate, 1,3,5(10)-Estratriene-3,17beta-diol Dipropionate, 3,17beta-Dihydroxy-1,3,5(10)-estratriene 3,17-Dipropionate, 17beta-Estradiol 3,17-Dipropionate

E1344-250MG E1344-1G
TCIAE1344-250MGEA 168 EUR
TCIAE1344-250MG TCIAE1344-1G
Estradiol Dipropionate ≥98.0% (by GC)
Estradiol Dipropionate
Formula: C₂₄H₃₂O₄
Peso molecolare: 384,52 g/mol
MDL Number: MFCD00051146
CAS Number: 113-38-2
EINECS: 204-026-5
Merck Index: 14,03703

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 105.0 to 109.0 deg-C
Specific rotation [a]20/D +38.0 to +42.0 deg (C=1, dioxane)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR