Stampa…

Ethylene Glycol bis(3-mercaptopropionate) ≥98.0% (by titrimetric analysis), purificata

Fornitore: TCI EUROPE
Attenzione

Synonyms: Ethylene bis(3-mercaptopropionate), Glycol bis(3-mercaptopropionate)

E1329-25G E1329-100G
TCIAE1329-25GEA 109 EUR
TCIAE1329-25G TCIAE1329-100G
Ethylene Glycol bis(3-mercaptopropionate) ≥98.0% (by titrimetric analysis), purificata
Ethylene Glycol bis(3-mercaptopropionate)
Formula: C₈H₁₄O₄S₂
Peso molecolare: 238,32 g/mol
Flash Pt: 255°C
MDL Number: MFCD00053520
CAS Number: 22504-50-3
EINECS: 245-044-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e91f1fc044508a2613adc08c5e1a598d1bde0c3&vid=5e91f1fc044508a2613adc08c5e1a598d1bde0c3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 96.0 %
Purity (Iodometric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR