Stampa…

Ethyl 2,2-Difluoro-2-(trimethylsilyl)acetate ≥97.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Ethyl 2,2-difluoro-2-(trimethylsilyl)acetate, 2,2-Difluoro-2-(trimethylsilyl)acetic acid Ethyl ester

E1314-1G E1314-5G
TCIAE1314-1GEA 69 EUR
TCIAE1314-1G TCIAE1314-5G
Ethyl 2,2-Difluoro-2-(trimethylsilyl)acetate ≥97.0% (by GC)
Ethyl 2,2-Difluoro-2-(trimethylsilyl)acetate
Formula: C₇H₁₄F₂O₂Si
Peso molecolare: 196,27 g/mol
Punto di ebollizione: 55°C
MDL Number: MFCD04973092
CAS Number: 205865-67-4
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=926e1403fdbb5887c47ba9e5595c702e5481b8c3&vid=926e1403fdbb5887c47ba9e5595c702e5481b8c3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid to cloudy liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR