Ethyl 6-[(2-Cyanoethoxy)(4,4-diphenylcyclohexyloxy)phosphoryloxy]hexanoate ≥95.0% (by HPLC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
6-[(2-Cyanoethoxy)(4,4-diphenylcyclohexyloxy)phosphoryloxy]hexanoic acid Ethyl ester
E1313-100MG
E1313-500MG
TCIAE1313-100MGEA
205
EUR
TCIAE1313-100MG
TCIAE1313-500MG
Ethyl 6-[(2-Cyanoethoxy)(4,4-diphenylcyclohexyloxy)phosphoryloxy]hexanoate ≥95.0% (by HPLC)
Ethyl 6-[(2-Cyanoethoxy)(4,4-diphenylcyclohexyloxy)phosphoryloxy]hexanoate
Formula:
C₂₉H₃₈NO₆P Peso molecolare: 527,60 g/mol Temperatura di stoccaggio: Refrigerata |
CAS Number:
572913-59-8 |
Risultati del test di specificazione
Purity (HPLC) | min. 95.0 area% |
Elemental analysis (Nitrogen) | 2.50 to 2.80 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...