Stampa…

Ethynyl(diphenyl)phosphine oxide ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

E1310-200MG E1310-1G
TCIAE1310-200MGEA 106 EUR
TCIAE1310-200MG TCIAE1310-1G
Ethynyl(diphenyl)phosphine oxide ≥98.0% (by GC)
Ethynyl(diphenyl)phosphine oxide
Formula: C₁₄H₁₁OP
Peso molecolare: 226,21 g/mol
CAS Number: 6104-48-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Brown powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR