Stampa…

1-Ethyl-3-methylimidazolium methylsulphate ≥98.0% (by HPLC)

Fornitore: TCI
Attenzione

Synonyms: 1-Ethyl-3-methyl-1H-imidazol-3-ium methyl sulphate

E1308-5G E1308-25G
TCIAE1308-5GEA 23 EUR
TCIAE1308-5G TCIAE1308-25G
1-Ethyl-3-methylimidazolium methylsulphate ≥98.0% (by HPLC)
1-Ethyl-3-methylimidazolium methylsulphate
Formula: C₇H₁₄N₂O₄S
Peso molecolare: 222.27 g/mol
Densità: 1.28
MDL Number: MFCD08457697
CAS Number: 516474-01-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f06f53ce2321735add3218ff67738db5ba782849&vid=f06f53ce2321735add3218ff67738db5ba782849&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white clear liquid
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR