1-Ethyl-3-methylimidazoliumtrifluoroacetate ≥97.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1-ethyl-3-methylimidazolium trifluoroacetate
E1307-5G
E1307-25G
TCIAE1307-5GEA
48
EUR
TCIAE1307-5G
TCIAE1307-25G
1-Ethyl-3-methylimidazoliumtrifluoroacetate ≥97.0% (by titrimetric analysis)
1-Ethyl-3-methylimidazoliumtrifluoroacetate
Formula:
C₈H₁₁F₃N₂O₂ Peso molecolare: 224,1804 g/mol |
MDL Number:
MFCD08458492 CAS Number: 174899-65-1 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (HPLC) | min. 93.0 area% |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...