(1-Ethoxycarbonylethyl)triphenylphosphonium bromide ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Synonyms:
(1-Ethoxy-1-oxopropan-2-yl)triphenylphosphonium bromide
Formula:
C₂₃H₂₄BrO₂P Peso molecolare: 443.33 g/mol |
MDL Number:
MFCD00031605 CAS Number: 30018-16-7 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...