Stampa…

2-(2-Ethylhexyl)thiophene ≥98.0% (by GC)

Fornitore: TCI

E1294-1ML E1294-5ML
TCIAE1294-1MLEA 68 EUR
TCIAE1294-1ML TCIAE1294-5ML
2-(2-Ethylhexyl)thiophene ≥98.0% (by GC)
2-(2-Ethylhexyl)thiophene
Formula: C₁₂H₂₀S
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD26743668
CAS Number: 4891-44-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32556fb944f82486cfc21005cf980fbb09bab9e4&vid=2bd1823b29c926d3ab4a5796612563f3bbc5fe28&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR