Stampa…

Etretinate ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Ethyl all-trans-9-(4-Methoxy-2,3,6-trimethylphenyl)-3,7-dimethyl-2,4,6,8-nonatetraenoate, all-trans-9-(4-Methoxy-2,3,6-trimethylphenyl)-3,7-dimethyl-2,4,6,8-nonatetraenoic acid Ethyl ester, Acitretin ethyl ester

E1293-50MG E1293-250MG
TCIAE1293-50MGEA 73 EUR
TCIAE1293-50MG TCIAE1293-250MG
Etretinate ≥98.0% (by HPLC)
Etretinate
Formula: C₂₃H₃₀O₃
Peso molecolare: 354,49 g/mol
MDL Number: MFCD00866624
CAS Number: 54350-48-0
EINECS: 259-119-3
UN: 3077
ADR: 9,III
Merck Index: 14,03892
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b60a80c37dc860ed8b521decb878913363f74245&vid=b60a80c37dc860ed8b521decb878913363f74245&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 103.0 to 107.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR